missing translation for 'onlineSavingsMsg'
Learn More

B Braun™ Mandrin for Vasofix™

Product Code. 10701314
Change view
Click to view available options
Unit Size:
Pack of 50
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. unitSize
10701314 Pack of 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10701314 Supplier B Braun™ Supplier No. 4219120

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Mandrin for Vasofix™ Ideal for interrupting infusion/transfusion

  • Usable for:
  • Vasofix™ Safety
  • Vasofix™ Braunüle™
  • Vasofix™ Certo
  • Safe closure of the lumen
  • Color size indicator
  • Identical with Vasofix™
  • Lock system
  • Latex-free
  • PVC-free

Specifications

Color Green
Diameter (Metric) Inner 18 mm
Latex Free Yes
Length (Metric) 45 mm
For Use With (Equipment) Vasofix Safety,Vasofix Braunüle, Vasofix Certo
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.