Learn More
Invitrogen™ MAN1C1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588794
Description
Immunogen sequence: PHSSRLKRLF LAPRTQQPGL EVVAEIAGHA PAREQEPPPN PAPAAPAPGE DDPSSWASPR RRKGGLRRTR PTGPREEATA ARGNSIPASR PGDEGVPFRF DFNAFRSRLR HPVLGTRADE SQEPQSQVRA QREKIKEMMQ FAWQSYKRYA MGKNELRPLT KDGYEGNMFG GLSGATVIDS LDTLYLMELK EEFQEAK; Positive Samples: A-549, U-87MG, HepG2, Mouse liver; Cellular Location: Golgi apparatus membrane, Single-pass type II membrane protein.
MAN1C1 gene ontology annotations related to this gene include metabolic process.
Specifications
| MAN1C1 | |
| Polyclonal | |
| Unconjugated | |
| MAN1C1 | |
| 1,2-alpha-mannosidase IC; AI593348; Alpha-1,2-mannosidase IC; HMIC; LOW QUALITY PROTEIN: mannosyl-oligosaccharide 1,2-alpha-mannosidase IC; MAN1A3; MAN1C; MAN1C1; Mannosidase alpha class 1C member 1; mannosidase, alpha, class 1C, member 1; mannosyl-oligosaccharide 1,2-alpha-mannosidase IC; mannosyl-oligosaccharide 1,2-alpha-mannosidase IC; LOW QUALITY PROTEIN: mannosyl-oligosaccharide 1,2-alpha-mannosidase IC; pp6318; processing alpha-1,2-mannosidase IC | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 230815, 57134 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 4.28 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| Q9NR34 | |
| MAN1C1 | |
| Recombinant protein (or fragment). | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.