missing translation for 'onlineSavingsMsg'
Learn More

MALT1 inhibitor MI-2, MedChemExpress

Product Code. 16425796
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16425796

Brand: Medchem Express HY12276

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress MALT1 inhibitor MI-2 is a MALT1 inhibitor (IC50=5.84 μM). MALT1 inhibitor MI-2 binds directly to MALT1, irreversibly suppresses protease function and is accompanied by NF-κB reporter activity suppression, c-REL nuclear localization inhibition, and NF-κB target gene downregulation. MALT1 inhibitor MI-2 shows nontoxic to animals.

MALT1 inhibitor MI-2 is a MALT1 inhibitor (IC50=5.84 μM). MALT1 inhibitor MI-2 binds directly to MALT1, irreversibly suppresses protease function and is accompanied by NF-κB reporter activity suppression, c-REL nuclear localization inhibition, and NF-κB target gene downregulation. MALT1 inhibitor MI-2 shows nontoxic to animals.

Specifications

Chemical Name or Material MALT1 inhibitor MI-2
CAS 1047953-91-2
Color White
Purity Grade Notes Research
Molecular Formula C19H17Cl3N4O3
Quantity 100 mg
Solubility Information DMSO : ≥ 46 mg/mL (100.94 mM)
SMILES O=C(NC1=CC=C(N2N=C(OCCOC)N=C2C3=CC=C(Cl)C(Cl)=C3)C=C1)CCl
Molecular Weight (g/mol) 455.72
Formula Weight 455.72
Percent Purity 98.08%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.