missing translation for 'onlineSavingsMsg'
Learn More
Learn More
B Braun™ Male Luer Lock Stopper
Description
- Made of polyethylene
- Sterile closings of all kinds of open IVs and IV sets, and pre-filled syringes
- White in color
- Provided in boxes of 100
Specifications
Specifications
| Material | Polyethylene |
| Color | White |
| Description | Male luer lock |
| For Use With (Application) | Sterile closings |
| Product Type | Closing Cone |
| Quantity | 100/Pk. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?