missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAK Monoclonal Antibody (OTI1G2)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527054
This item is not returnable.
View return policy
Description
MAK Monoclonal Antibody for Western Blot, IHC (P)
The product of this gene is a serine/threonine protein kinase related to kinases involved in cell cycle regulation. It is expressed almost exclusively in the testis, primarily in germ cells. Studies of the mouse and rat homologs have localized the kinase to the chromosomes during meiosis in spermatogenesis, specifically to the synaptonemal complex that exists while homologous chromosomes are paired. There is, however, a study of the mouse homolog that has identified high levels of expression in developing sensory epithelia so its function may be more generalized.
Specifications
| MAK | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P20794 | |
| MAK | |
| Human recombinant protein fragment corresponding to amino acids 291-623 of MAK produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1G2 | |
| Unconjugated | |
| MAK | |
| A930010O05Rik; dJ417M14.2; MAK; male germ cell associated kinase; male germ cell-associated kinase; male germ cell-associated kinase retinal-enriched isoform; protein kinase RCK; Rck; RP62; serine/threonine protein kinase MAK; serine/threonine-protein kinase MAK; testicular secretory protein Li 28; Unknown (protein for MGC:134523) | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4117 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction