Learn More
Magnolin, TRC
Specifications
Specifications
| Chemical Name or Material | (+)-Magnolin |
| CAS | 31008-18-1 |
| Molecular Formula | C23H28O7 |
| Quantity | 25 mg |
| Synonym | 1H,3H-Furo[3,4-c]furan, 1-(3,4-dimethoxyphenyl)tetrahydro-4-(3,4,5-trimethoxyphenyl)-, (1S,3aR,4S,6aR)-, 1H,3H-Furo[3,4-c]furan, 1-(3,4-dimethoxyphenyl)tetrahydro-4-(3,4,5-trimethoxyphenyl)-, [1S-(1α,3aα,4α,6aα)]-, 1H,3H-Furo[3,4-c]furan, 1α-(3,4-dimethoxyphenyl)-3aα,4,6,6aα-tetrahydro-4α-(3,4,5-trimethoxyphenyl)-, (+)- (8CI), (+)-Magnolin, (1S,3aR,4S,6aR)-1-(3,4-Dimethoxyphenyl)tetrahydro-4-(3,4,5-trimethoxyphenyl)-1H,3H-furo[3,4-c]furan, Magnolin, Medioresinol dimethyl ether |
| InChI Formula | InChI=1S/C23H28O7/c1-24-17-7-6-13(8-18(17)25-2)21-15-11-30-22(16(15)12-29-21)14-9-19(26-3)23(28-5)20(10-14)27-4/h6-10,15-16,21-22H,11-12H2,1-5H3/t15-,16-,21+,22+/m0/s1 |
| SMILES | COc1ccc(cc1OC)[C@H]2OC[C@H]3[C@@H]2CO[C@@H]3c4cc(OC)c(OC)c(OC)c4 |
| IUPAC Name | (3S,3aR,6S,6aR)-3-(3,4-dimethoxyphenyl)-6-(3,4,5-trimethoxyphenyl)-1,3,3a,4,6,6a-hexahydrofuro[3,4-c]furan |
| Molecular Weight (g/mol) | 416.46 |
| Formula Weight | 416.1835 |