Learn More
Invitrogen™ MagniSort™ Mouse F4/80 Positive Selection Kit
Click to view available options
Quantity:
200 tests
Unit Size:
200 tests
Description
Includes
- MagniSort™ Anti-Mouse F4/80 Biotin : 200 tests, 20μL/test
- MagniSort™ Positive Selection Beads B
- F4/80+ cells are bound by antibody and then magnetic beads
- When placed in a magnetic field, the undesired cells can be separated from F4/80+ cells by decanting
- After positive selection, the purity of selected cells can be verified by staining with Anti-Mouse F4/80, clone BM8 and Anti-Mouse CD11b, clones M1/70
- To reduce non-specific binding, the MagniSort™ Anti-Mouse F4/80 Biotin contains Fc Receptor block
Specifications
Specifications
| Cell Type | F4/80+ |
| Product Type | Positive Selection Kit |
| No. of Tests | 200 tests |
| Quantity | 200 tests |
| Target Species | Mouse |