Learn More
Invitrogen™ MagniSort™ Human CD3 Positive Selection Kit
Product Code. 15541247
Click to view available options
Quantity:
200 tests
Unit Size:
200 tests
Contains Sodium azide
Product Code. 15541247
Brand: Invitrogen™ 8802683074
Please to purchase this item. Need a web account? Register with us today!
Contains Sodium azide
Includes
  • MagniSort Anti-Human CD3 Biotin : 200 tests, 20μL/test
  • MagniSort Positive Selection Beads A
Designed for the magnetic separation of CD3+ cells by positive selection. It has been optimized for the isolation of CD3+ cells from human peripheral blood monocuclear cells, utilizing a biotinylated Anti-Human CD3 antibody and streptavidin-coated magnetic beads.
  • CD3+ cells are bound by antibody and then magnetic beads
  • When placed in a magnetic field, the undesired cells can be separated from CD3+ cells by decanting
  • After positive selection, the purity of selected cells can be verified by staining with Anti-Human CD3, clones UCHT1 or SK7
TRUSTED_SUSTAINABILITY
Specifications
Cell Type CD3+
Product Type Positive Selection Kit
No. of Tests 200 tests
Quantity 200 tests
Target Species Human