Learn More
Buerkle™ Magnetic Stirring Bar
Product Code. 10184532 Shop All Bürkle Products
Click to view available options
Length (Metric):
20 mm
25 mm
30 mm
40 mm
50 mm
80 mm
Diameter (Metric):
6 mm
7 mm
8 mm
10 mm
Unit Size:
Each
Product Code. 10184532
Brand: Buerkle™ 82712507
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Molded with an outer layer of chemically inert PTFE for durability and non-reactivity. Buerkle™ Magnetic Stirring Bars feature a cylindrical shape and smooth surface for excellent mixing capability.
  • Opaque PTFE construction
  • High performance permanent magnets 
  • Long lifespan
  • Exceptional temperature resistance
  • Wide range of sizes
  • Stirring bars with diameters of 6 and 8mm can be used with Buerkle™ Fish-clip™ for extra gentle stirring
TRUSTED_SUSTAINABILITY
Specifications
Color White
Diameter (Metric) 7 mm
Length (Metric) 25 mm
Shape Cylinder