missing translation for 'onlineSavingsMsg'
Learn More

Magnesium hydrogen phosphate trihydrate, 97+%

Product Code. 11442437
Click to view available options
Quantity:
250 g
1000 g
Unit Size:
1000g
250g
This item is not returnable. View return policy

Product Code. 11442437

Brand: Thermo Scientific Alfa Aesar A14279.0B

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7782-75-4 | H7MgO7P | 174.33 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-75-4
Molecular Formula H7MgO7P
Molecular Weight (g/mol) 174.33
MDL Number MFCD00150390
InChI Key OKIWLDVQGKRUNR-UHFFFAOYSA-L
Synonym magnesium hydrogen phosphate trihydrate, dimagnesium phosphate, unii-hf539g9l3q, magnesium phosphate dibasic trihydrate, magnesium 2+ trihydrate hydrogen phosphate, magnesium 2+ ion trihydrate hydrogen phosphate, kc defense, newberyite, magnesium dibasic phosphate trihydrate, acmc-20aklg
PubChem CID 16204530
IUPAC Name magnesium(2+) trihydrate hydrogen phosphate
SMILES O.O.O.[Mg++].OP([O-])([O-])=O

Specifications

Quantity 1000 g
Merck Index 14,5681
Formula Weight 174.34 (120.28 Anhydrous)
Percent Purity 99%
Odor Odorless
Density 2.13
Chemical Name or Material Magnesium hydrogen phosphate trihydrate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.