missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Magnesium acetate tetrahydrate, for biochemistry
Click to view available options
Quantity:
250 g
1 kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For biochemistry
Specifications
Melting Point | 80.0°C |
Quantity | 250 g |
Linear Formula | (CH3CO2)2Mg·4H2O |
Fieser | 15,196 |
Merck Index | 15,5718 |
Solubility Information | Solubility in water: 1200g/L (15°C). Other solubilities: soluble in alcohol |
Formula Weight | 214.46 |
Grade | Biochemical |
Packaging | Plastic bottle |
Chemical Name or Material | Magnesium acetate tetrahydrate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'