missing translation for 'onlineSavingsMsg'
Learn More

Applied Biosystems™ MagMAX™ TFS Solution

Product Code. 30054766 Shop All Applied Biosystems Products
Change view
Click to view available options
Quantity:
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30054766 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30054766 Supplier Applied Biosystems™ Supplier No. A45351

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MagMAX™ TFS Solution is a tissue fragment stability and storage solution developed to ensure the stability of RNA/DNA/proteins during room temperature transportation and/or storage

The MagMAX™ TFS Solution was developed for use with ear tissue biopsies. Applications include genomic DNA sequencing (including CE, microarray, and next-generation sequencing), RNA detection with RT-PCR, and ELISA.

TRUSTED_SUSTAINABILITY

Specifications

Color Clear
pH 7.4 ±0.05
Quantity 1 L

For Veterinary Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.