Learn More
Description
MagMAX™ CORE Glass Microbeads aid in the homogenization of completely dessicated feces such as environmental samples or fecal pellets.
For a kit that contains these microbeads as well as the MagMAX™ CORE Nucleic Acid Purification Kit and the MagMAX™ CORE Mechanical Lysis Module, please see Cat. No. A37488.
Specifications
Specifications
| Content And Storage | 50 mL bottle, store at room temperature |
| Sample Type | Blood, Buccal Samples, Cells, DNA, RNA, Tissue, Virus |
| For Use With (Application) | Next-Generation Sequencing, RT-PCR (Endpoint), Real-Time PCR |
| High-throughput Compatibility | Manual Protocols, Multiplexing |
| Product Line | MagMAX |
| Quantity | 100 Tests |
| Shipping Condition | Room Temperature |
| Target | Total RNA, Pathogen DNA/RNA, Genomic DNA, Viral DNA/RNA |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.