missing translation for 'onlineSavingsMsg'
Learn More

Brady Magicard™ Prima 8 - Clear Laminate Patch Film

Product Code. 30067323
Click to view available options
Color:
Transparent
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 30067323

Brand: Brady 321993

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Prima 8 - Clear laminate patch film.

  • Provide extra protection against scratches on your card.
  • 0.6 mil thick laminate.
  • Can be applied with Prima 8 Laminator.

Specifications

Product Type Laminate
Color Transparent
For Use With (Equipment) Prima 8 Laminator
Print Technology Thermal Transfer
Description Clear Patch Laminate 0.6mil for Prima 8 Laminator
Format Laminate
Includes 1 Laminate Roll
For Use With (Application) Healthcare and Hospital labelling;General identification;Facility identification;Security identification
Quantity 1 Ea.
Alternate Part Number PRIMA462
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.