missing translation for 'onlineSavingsMsg'
Learn More

Brady Magicard™ 300/600 - Cleaning Kit

Product Code. 30067312
Click to view available options
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 30067312

Brand: Brady 321983

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Cleaning kit for the Magicard 300 or Magicard 600.

  • Ensure high quality printing on every card.
  • Good for 10 cleaning cycles.
  • Kit holds cleaning cards and cleaning pen.

Specifications

Product Type Cleaning Kit
For Use With (Equipment) Magicard 300, Magicard 600
Description Cleaning Kit 10 Cards, 1 Pen for Magicard 300/600 Printer
Format Cleaning Kit
Includes 10 Cleaning Cards, 1 Cleaning Pen
For Use With (Application) Healthcare and Hospital labelling;General identification;Facility identification;Security identification
Quantity 1 Ea.
Alternate Part Number 3633-0053
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.