missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MAGEB6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5140289
This item is not returnable.
View return policy
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: SSSSSSRACL GDCRRSSDAS IPQESQGVSP TGSPDAVVSY SKSDVAANGQ DEKSPSTSRD.
This gene is a member of the MAGEB gene family. The members of this family have their entire coding sequences located in the last exon, and the encoded proteins show 50 to 68% sequence identity to each other. The promoters and first exons of the MAGEB genes show considerable variability, suggesting that the existence of this gene family enables the same function to be expressed under different transcriptional controls. This gene is expressed in testis, and in a significant fraction of tumors of various histological types. The MAGEB genes are clustered on chromosome Xp22-p21.
Specifications
| MAGEB6 | |
| Polyclonal | |
| Unconjugated | |
| MAGEB6 | |
| cancer/testis antigen 3.4; cancer/testis antigen family 3, member 4; CT3.4; MAGE family member B6; MAGEB6; MAGE-B6; MAGE-B6 antigen; MAGEB6A; melanoma antigen family B, 6; melanoma antigen family B6; melanoma-associated antigen B6 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 158809 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8N7X4 | |
| MAGEB6 | |
| Recombinant protein corresponding to Human MAGEB6. Recombinant protein control fragment (Product #RP-109586). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction