missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MAF1 Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00084232-B01P
This item is not returnable.
View return policy
Description
MAF1 Polyclonal antibody specifically detects MAF1 in Human samples. It is validated for Western Blot
Specifications
| MAF1 | |
| Polyclonal | |
| PBS (pH 7.4) | |
| DKFZp586G1123, homolog of yeast MAF1, MAF1 homolog (S. cerevisiae), MGC20332, MGC31779, MGC39758, repressor of RNA polymerase III transcription MAF1 homolog | |
| MAF1 (AAH31273.1, 1 a.a. - 256 a.a.) full-length human protein. MKLLENSSFEAINSQLTVETGDAHIIGRIESYSCKMAGDDKHMFKQFCQEGQPHVLEALSPPQTSGLSPSRLSKSQGGEEEGPLSDKCSRKTLFYLIATLNESFRPDYDFSTARSHEFSREPSLSWVVNAVNCSLFSAVREDFKDLKPQLWNAVDEEICLAECDIYSYNPDLDSDPFGEDGSLWSFNYFFYNKRLKRIVFFSCRSISGSTYTPSEAGNELDMELGEEEVEEESRSRGSGAEETSTMEEDRVPVICI | |
| 50 μg | |
| DNA replication Transcription Translation and Splicing | |
| 84232 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| AAH31273.1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction