missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macrophage Rat anti-Mouse, Clone: MOMA-2, Novus Biologicals™
Rat Monoclonal Antibody
668.00 €
Specifications
| Antigen | Macrophage |
|---|---|
| Clone | MOMA-2 |
| Concentration | 0.5 mg/ml |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
Beskrivelse
Macrophage Monoclonal specifically detects Macrophage in Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Frozen, Knockout Validated.Tekniske data
| Macrophage | |
| 0.5 mg/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS | |
| Mouse lymph node stroma | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| MOMA-2 | |
| Flow Cytometry | |
| Unconjugated | |
| Rat | |
| Mouse | |
| EC 5.3.2.1, EC 5.3.3.12, GIFmacrophage migration inhibitory factor, GLIF, Glycosylation-inhibiting factor, L-dopachrome isomerase, L-dopachrome tautomerase, macrophage migration inhibitory factor (glycosylation-inhibiting factor), MMIF, Phenylpyruvate tautomerase | |
| IgG2b | |
| Protein G purified |
Ser du en mulighed for forbedring?Del en indholdskorrektion
Korrektion af produktindhold
Dit input er vigtigt for os. Udfyld denne formular for at give feedback relateret til indholdet på dette produkt.
Produkttitel