missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ MacConkey Broth Purple with Durham Tube
Description
Selectively detect and enumerate coliform bacteria in food, milk or water samples by observing lactose fermentation (colour change) and gas production, using MacConkey Broth Purple with Durham Tube
Specifications
Specifications
| Color | Purple |
| Content And Storage | 2°C to 25°C |
| Format | Universal bottle |
| Manufacturing Origin | United Kingdom |
| Product Type | Microbiology Media |
| Fill Weight | 10.0 to 10.3 g |
| Packaging Type | 24 Universal Bottles with screw cap closures in a box |
| Quantity | 24/Pk. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?