Learn More
m-Toluenesulfonic acid monohydrate, 97%
Product Code. 11362146
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11362146 25 g 25g
11352146 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11362146 Supplier Thermo Scientific Alfa Aesar Supplier No. L03419.14
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
CAS: 312619-56-0 | C7H10O4S | 190.213 g/mol
m-Toluenesulfonic acid monohydrate is used in chemical research.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
m-Toluenesulfonic acid monohydrate is used in chemical research.
Solubility
Fully miscible in water.
Notes
Hygroscopic. Store away from strong bases, moisture. Incompatible materials are oxidizing agents, bases.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 312619-56-0
Molecular Formula C7H10O4S
Molecular Weight (g/mol) 190.213
MDL Number MFCD00167130
InChI Key GHLYGGIKYFLYTH-UHFFFAOYSA-N
Synonym 3-methylbenzenesulfonic acid hydrate, m-toluenesulfonic acid monohydrate, m-toluenesulfonic acid hydrate, acmc-20akdl, m-toluenesulfonic acid 1-hydrate, 3-toluenesulfonic acid 1-hydrate, 3-methylbenzenesulfonic acid hydrate 1:x, benzenesulfonicacid, 3-methyl-, hydrate 1:1
PubChem CID 53249652
IUPAC Name 3-methylbenzenesulfonic acid;hydrate
SMILES CC1=CC(=CC=C1)S(=O)(=O)O.O
Specifications
Boiling Point 165°C (0.1 mmHg)
Refractive Index 1.527
Quantity 25 g
UN Number UN2586
Beilstein 2208421
Solubility Information Fully miscible in water.
Formula Weight 190.22 (172.20 Anhydrous)
Percent Purity 97%
Chemical Name or Material m-Toluenesulfonic acid monohydrate