missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ M 084 hydrochloride

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
M 084 hydrochloride is a TRPC4/5 channel blocker (IC50 values are 3.7-10.3 and 8.2 μM, respectively). Also weakly blocks TRPC3 channels. Exhibits rapid antidepressant and anxiolytic effects in vivo.
Specifications
Specifications
| Chemical Name or Material | N-Butyl-1H-benzimidazol-2-amine hydrochloride |
| CAS | 1992047-63-8 |
| Quantity | 10 mg |
| Target | TRPC Channel Blockers |
| Molecular Formula | C11H15N3.HCl |
| Formula Weight | 225.72 |
| Purity | >99% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction