missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LYST Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
280.00 € - 589.00 €
Specifications
| Antigen | LYST |
|---|---|
| Applications | Immunocytochemistry, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18266411
|
Novus Biologicals
NBP2-58768 |
100 μL |
589.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18623297
|
Novus Biologicals
NBP2-58768-25ul |
25 μL |
280.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LYST Polyclonal specifically detects LYST in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.Specifications
| LYST | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 1130 | |
| This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:KKYETEEGVNKAAWQKTVNNNQQSLFQRLDSKSKDISKIAADITQAVSLSQGNERKKVIQHIRGMYKVDLSASRHWQELIQQLTHDRAVWYDPIYYPTSWQ | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Immunocytochemistry, Immunofluorescence | |
| Unconjugated | |
| RUO | |
| Beige homolog, Chediak-Higashi syndrome 1, CHSCHS1beige protein, lysosomal trafficking regulator, lysosomal-trafficking regulator | |
| LYST | |
| IgG | |
| Affinity Purified |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title