Learn More
Lyral, mixture of isomers, TRC
Product Code. 17963085
Change view
Click to view available options
Quantity:
100 mg
1 g
10 g
Unit Size:
100mg
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17963085 1 g 1g
17962225 10 g 10g
17900435 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 17963085 Supplier Toronto Research Chemicals Supplier No. TRCL4878701G
Please to purchase this item. Need a web account? Register with us today!
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 31906-04-4
Molecular Formula C13 H22 O2
Molecular Weight (g/mol) 210.31
Synonym 4-(4-Hydroxy-4-methylpentyl)-3-cyclohexene-1-carboxaldehyde, 4-(4-Hydroxy-4-methylpentyl)-3-cyclohexenecarboxaldehyde, 4-(4-Methyl-4-hydroxyamyl)cyclohex-3-ene carboxaldehyde, Landolal, Liral
IUPAC Name 4-(4-hydroxy-4-methylpentyl)cyclohex-3-ene-1-carbaldehyde
SMILES CC(C)(O)CCCC1=CCC(CC1)C=O
Specifications
Quantity 1 g
InChI Formula InChI=1S/C13H22O2/c1-13(2,15)9-3-4-11-5-7-12(10-14)8-6-11/h5,10,12,15H,3-4,6-9H2,1-2H3
Formula Weight 210.162
Chemical Name or Material Lyral (mixture of isomers)