Learn More
Lycopene, MedChemExpress
Product Code. p-200098861
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16458938 10 mg 10mg
16454807 50 mg 50mg
16431378 5 mg 5mg
16413856 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16458938 Supplier Medchem Express Supplier No. HYN0287
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Lycopene is naturally occurring carotenoids found in tomato, tomato products, and in other red fruits and vegetables; exhibits antioxidant effects.
Lycopene is naturally occurring carotenoids found in tomato, tomato products, and in other red fruits and vegetables
Specifications
Chemical Name or Material Lycopene
CAS 502-65-8
Color Reddish Brown
Purity Grade Notes Research
Molecular Formula C40H56
Quantity 10 mg
Solubility Information THF : 5 mg/mL (9.31 mM; Need ultrasonic) ∣Ethanol : < 1 mg/mL (ultrasonic) (insoluble)
SMILES C/C(C)=C\CC/C(C)=C/C=C/C(C)=C/C=C/C(C)=C/C=C/C=C(/C=C/C=C(C)/C=C/C=C(CC/C=C(C)\C)\C)C
Molecular Weight (g/mol) 536.87
Formula Weight 536.87
Percent Purity 98.0%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation