missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies LY411575

Product Code. 17170122
Encompass_Preferred
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 17170122

Brand: STEMCELL Technologies 72792

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Notch pathway inhibitor; Inhibits γ-secretase

  • LY411575 is a cell-permeable γ-secretase inhibitor (IC50 = 0.14 nM) that blocks Notch activation in vitro at 500 μM (Curry et al.; Czirr et al.) via γ-Secretase inhibition
  • Notch is a transmembrane receptor that plays a key role in cell fate decisions including cell proliferation, differentiation, and apoptosis
  • Promotes neuronal differentiation of neural progenitor cells derived from mouse embryonic stem (ES) cells (Abranches et al.; Aranha et al.)
  • Promotes goblet cell differentiation in mouse intestine and cultured colonic organoids (Okamoto et al.; Yui et al.)
  • Induces hair cell differentiation from inner ear stem cells in vitro, and transdifferentiation of supporting cells into hair cells in vivo (Bramhall et al.; Mizutari et al.)
  • Causes premature differentiation of Her4-positive progenitors into neurons in zebrafish (Dirian et al.)
  • Induces apoptosis in primary and immortalized Karposi's sarcoma cells (Curry et al.)

Specifications

CAS 209984-57-6
Quantity 5 mg
Molecular Formula C26H23F2N3O4
Purity ≥98%
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.