Learn More
LY3200882, MedChemExpress
Product Code. 16416507
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16416507 5 mg 5mg
16480157 50 mg 50mg
16473778 10 mg 10mg
16454836 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 16416507 Supplier Medchem Express Supplier No. HY103021
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress LY3200882 is a potent, highly selective, ATP-competitive and orally active TGF-β receptor type 1 (ALK5) inhibitor with an IC50 of 38.2 nM. LY3200882 inhibits various pro-tumorigenic activities and is also used as an immune modulatory agent.
LY3200882 is a potent, highly selective, ATP-competitive and orally active TGF-β receptor type 1 (ALK5) inhibitor with an IC50 of 38.2 nM. LY3200882 inhibits various pro-tumorigenic activities and is also used as an immune modulatory agent.
Specifications
Chemical Name or Material LY3200882
CAS 1898283-02-7
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C24H29N5O3
Quantity 5 mg
Solubility Information DMSO : 33.33 mg/mL (76.53 mM; Need ultrasonic)
SMILES OC(C)(C)C1=NC=CC(NC2=NC=CC(OC3=CN(C4CC4)N=C3C5CCOCC5)=C2)=C1
Molecular Weight (g/mol) 435.52
Formula Weight 435.52
Percent Purity 98.82%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease