Learn More
LY2119620, MedChemExpress
Product Code. 16489427
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16489427
Brand: Medchem Express HY15885
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress LY2119620 is a high-affinity muscarinic M2/M4 receptor agonist.
LY2119620 is a high-affinity muscarinic M2/M4 receptor agonist.
Specifications
Chemical Name or Material LY2119620
CAS 886047-22-9
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C19H24ClN5O3S
Quantity 5 mg
Solubility Information DMSO : 67.5 mg/mL (154.13 mM; Need ultrasonic)
SMILES NC1=C(C(NC2CC2)=O)SC3=C1C(C)=C(Cl)C(OCC(N4CCN(C)CC4)=O)=N3
Molecular Weight (g/mol) 437.94
Formula Weight 437.94
Percent Purity 98.77%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation