Learn More
Ly-6A/E Rat anti-Mouse, Brilliant Violet 786, Clone: E13-161.7, BD Optibuild™
Rat Monoclonal Antibody
Brand: BD Biosciences 744327
Description
The E13-161.7 mAb recognizes Ly-6A.2 and Ly-6E.1, which are allelic members of the Ly-6 multigene family. Ly-6A/E is also known as stem cell antigen 1 (Sca-1/Sca1), or T-cell-activating protein (TAP). Ly-6A/E is a phosphatidylinositol-anchored protein of ∽18kDa that is expressed on multipotent hematopoietic stem cells (HSC) in mice with both Ly-6 haplotypes. Sca-1+ HSC are found in the adult bone marrow and fetal liver, but not in the early embryo yolk sac or intraembryonic hematopoietic sites, and can be mobilized to the peripheral blood and spleen in the adult. In mice expressing the Ly-6.2 haplotype, Ly-6A/E is also expressed on distinct subpopulations of bone marrow and peripheral B lymphocytes, myeloid cells, and thymic and peripheral T lymphocytes, on the earliest intrathymic T-cell precursor population, and in several non-hematopoietic tissues. Strains with the Ly-6.1 haplotype have few Ly-6A/E+ resting peripheral lymphocytes, whereas activation of T cells from mice of both Ly-6 haplotypes leads to strong expression of the Sca-1 antigen. Studies with the D7 antibody have demonstrated that Ly-6A/E may be involved in the regulation of B and T lymphocyte responses, and it appears to be required for T-cell receptor-mediated T-cell activation. Purified E13-161.7 mAb can block binding of FITC-conjugated D7 antibody (anti-Ly-6A/E) to mouse splenocytes, but purified mAb D7 is unable to block binding of FITC-conjugated E13-161.7 antibody.
The antibody was conjugated to BD Horizon™ BV786 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 786nm. BD Horizon BV786 can be excited by the violet laser and detected in a filter used to detect Cy™7-like dyes (eg, 780/60nm filter). BV786 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 786 nm. BV786 offers a bright choice for the 6th detector off the violet laser.
Specifications
| Ly-6A/E | |
| E13-161.7 | |
| Brilliant Violet 786 | |
| SCA-1, Sca1, Stem cell antigen 1, TAP, T-cell-activating protein, Ly6 | |
| BALB/c mouse-derived “pre-T” cell hybridoma | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Rat | |
| Affinity Purified | |
| RUO | |
| Mouse | |
| IgG2a κ |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.