missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ LY 341495

Product Code. 30001209
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
This item is not returnable. View return policy

Product Code. 30001209

Brand: Tocris Bioscience™ 1209/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Highly potent and selective group II mGlu antagonist

Highly potent and selective group II metabotropic glutamate receptor antagonist (Ki/IC50 values are 2.3, 1.3, 173, 990, 6800, 8200 and 22000 nM for human mGlu2, mGlu3, mGlu8, mGlu7a, mGlu1a, mGlu5a and mGlu4a receptors respectively). Readily brain penetrant and active in vivo.

Disodium salt (Cat. No. 4062) also available.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 201943-63-7
Molecular Formula C20H19NO5

Specifications

Quantity 10 mg
Formula Weight 353.37
Purity >98%
Molecular Weight (g/mol) 353.374
Chemical Name or Material LY 341495
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.