missing translation for 'onlineSavingsMsg'
Learn More

LY 3000328, MedChemExpress

Product Code. 16436608
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16436608

Brand: Medchem Express HY15533

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress LY 3000328 is a potent and selective Cathepsin S (Cat S) inhibitor with IC50s of 7.7 and 1.67 nM for hCat S and mCat S, respectively.

LY 3000328 is a potent and selective Cathepsin S (Cat S) inhibitor with IC50s of 7.7 and 1.67 nM for hCat S and mCat S, respectively.

Specifications

Chemical Name or Material LY 3000328
CAS 1373215-15-6
Color White
Purity Grade Notes Research
Molecular Formula C25H29FN4O5
Quantity 10 mg
Solubility Information DMSO : ≥ 50 mg/mL (103.19 mM)
SMILES O=C(C1=CC=C(F)C=C1)N[C@@H]2[C@@H](OC(NC)=O)COC3=CC=C(N4CCN(C5COC5)CC4)C=C32
Molecular Weight (g/mol) 484.52
Formula Weight 484.52
Percent Purity 98.12%
Grade Research
Physical Form Solid
For Use With (Application) COVID-19-immunoregulation
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.