missing translation for 'onlineSavingsMsg'
Learn More
Learn More
LXR alpha/NR1H3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 5 publications
213.00 €
Specifications
| Antigen | LXR alpha/NR1H3 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:300, Flow Cytometry 3-5 μg/106 cells, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:50-1:500, Gel Super Shift Assays 1:1 - 1:100 |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
LXR alpha/NR1H3 Polyclonal specifically detects LXR alpha/NR1H3 in Human, Mouse samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Gel Super Shift Assays.Specifications
| LXR alpha/NR1H3 | |
| Western Blot 1:300, Flow Cytometry 3-5 μg/106 cells, Immunohistochemistry 1:10-1:500, Immunocytochemistry/Immunofluorescence 1:50-1:500, Gel Super Shift Assays 1:1 - 1:100 | |
| Polyclonal | |
| Rabbit | |
| Diabetes Research, Lipid and Metabolism | |
| PBS, 1 mg/ml BSA with 0.05% Sodium Azide | |
| 10062 | |
| Synthetic peptide corresponding to residues C D(318) F S Y N R E D F A K A(329) of human LXR alpha. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| alpha, Liver X receptor alpha, liver X receptor-alpha, LXRA, Nuclear receptor subfamily 1 group H member 3, nuclear receptor subfamily 1, group H, member 3, oxysterols receptor LXR-alpha, RLD-1 | |
| NR1H3 | |
| IgG | |
| Affinity Purified | |
| Detects recombinant human LXR alpha. This does not detect recombinant human LXR beta. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title