Learn More
LXR-623, MedChemExpress
Product Code. 16494078
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16494078 10 mg 10mg
16440437 50 mg 50mg
16425056 100 mg 100mg
16416787 5 mg 5mg
16401006 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 16494078 Supplier Medchem Express Supplier No. HY10629
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress LXR-623 is a brain-penetrant partial LXRα and full LXRβ agonist, with IC50s of 24 nM and 179 nM, respectively.
LXR-623 is a brain-penetrant partial LXRα and full LXRβ agonist, with IC50s of 24 nM and 179 nM, respectively.
Specifications
Chemical Name or Material LXR-623
CAS 875787-07-8
Color White
Purity Grade Notes Research
Molecular Formula C21H12ClF5N2
Quantity 10 mg
Synonym WAY 252623
Solubility Information DMSO : ≥ 47 mg/mL (111.17 mM)
SMILES ClC1=C(CN2N=C(C(C(F)(F)F)=CC=C3)C3=C2C4=CC=C(F)C=C4)C=CC(F)=C1
Molecular Weight (g/mol) 422.78
Formula Weight 422.78
Percent Purity 99.4%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death