missing translation for 'onlineSavingsMsg'
Learn More

LXR-623, MedChemExpress

Product Code. 16416787
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
200 mg
Unit Size:
100mg
10mg
200mg
50mg
5mg
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16416787 5 mg 5mg
16494078 10 mg 10mg
16440437 50 mg 50mg
16425056 100 mg 100mg
16401006 200 mg 200mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 16416787 Supplier Medchem Express Supplier No. HY10629

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress LXR-623 is a brain-penetrant partial LXRα and full LXRβ agonist, with IC50s of 24 nM and 179 nM, respectively.

LXR-623 is a brain-penetrant partial LXRα and full LXRβ agonist, with IC50s of 24 nM and 179 nM, respectively.

Specifications

Chemical Name or Material LXR-623
CAS 875787-07-8
Color White
Purity Grade Notes Research
Molecular Formula C21H12ClF5N2
Quantity 5 mg
Synonym WAY 252623
Solubility Information DMSO : ≥ 47 mg/mL (111.17 mM)
SMILES ClC1=C(CN2N=C(C(C(F)(F)F)=CC=C3)C3=C2C4=CC=C(F)C=C4)C=CC(F)=C1
Molecular Weight (g/mol) 422.78
Formula Weight 422.78
Percent Purity 99.4%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.