Learn More
LXN Mouse anti-Human, Clone: OTI1E10, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503779S
Description
LXN Monoclonal antibody specifically detects LXN in Human samples. It is validated for Flow Cytometry, Immunohistochemistry (Paraffin), Western BlotSpecifications
| LXN | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BS40 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 56925 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| OTI1E10 | |
| Unconjugated | |
| LXN | |
| ECI; endogenous carboxypeptidase inhibitor; Latexin; latexin protein; Lxn; MUM; Protein MUM; TCI; tissue carboxypeptidase inhibitor | |
| Full length human recombinant protein of human LXN produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.