Learn More
Greiner Bio-One LUMITRAC™ 200 HiBase μClear™-Bottom 1536-well Microplate
GREENER_CHOICE
Product Code. 13027963
Click to view available options
Color:
Clear
Unit Size:
Pack of 60
Product Code. 13027963
Brand: Greiner Bio-One 782095
Only null left
Add to basket
Add to basket
Free of detectable DNase, RNase, human DNA and pyrogens
Alphanumeric well coding
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance Free of detectable DNase/RNase, human DNA and pyrogens
Format Standard
Lid With Lid
Array 32 x 48
Autoclavable Not Autoclavable
Binding Type Medium Binding
Color Clear
Material Polystyrene
No. of Wells 1536
Volume (Metric) Working 3 to 10 μL
Sterility Non-sterile
Total Volume (Metric) Well 12.6 μL
Well Shape Rounded Square
Dnase Free Yes
Max. RCF 1000 x g (swinging-bucket rotor)
No. per Case 50
No. per Pack 16
RNase Free Yes
Temperature Range (Metric) -20°C to +60°C
Dimensions (L x W x H) 128 x 85 x 10 mm