Learn More
Poulten & Graf GmbH Sanitex glass and metal syringe
Description
Made of metal and glass
Amber graduations
Specifications
Specifications
| Autoclavable | Autoclavable |
| Material | Glass, Metal |
| Product Type | Syringe |
| Max. Temperature (Metric) | 134°C |
| Quantity | 1/Ea. |
| Volume (Metric) | 20 mL |