Learn More
MedChemExpress Luciferase
Product Code. 16499488
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
Product Code. 16499488
Brand: MedChemExpress HYP1004
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Luciferase from Vibrio fischeri has also been used in a study to investigate the sensitivity of dark mutants of various strains of luminescent bacteria to reactive oxygen species.
Specifications
Quantity 10 mg
Storage Requirements Powder -80°C 2 years, -20°C 1 year ∣ In solvent -80°C 6 months, -20°C 1 month
Solubility H2O : 50 mg/mL (Need ultrasonic)
Shipping Condition Room temperature
CAS 9014-00-0
Target NO Synthase ∣ MMP ∣ Endogenous Metabolite