missing translation for 'onlineSavingsMsg'
Learn More

Lubiprostone, MedChemExpress

Product Code. 16418578
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
10 mg
50 mg
Unit Size:
10mg
1mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16418578

Brand: Medchem Express HYB0679

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Lubiprostone(SPI-0211;RU0211) is a gastrointestinal agent used for the treatment of idiopathic chronic constipation.

Lubiprostone(SPI-0211

Specifications

Chemical Name or Material Lubiprostone
Physical Form Solid
CAS 136790-76-6
Color White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C20H32F2O5
For Use With (Application) Metabolism-sugar/lipid metabolism
Synonym RU-0211 SPI-0211
Solubility Information DMSO : 100 mg/mL (256.11 mM; Need ultrasonic)
SMILES O=C(C(F)(F)CCCC)CC[C@@H]([C@@H](C1)O)[C@H](C1=O)CCCCCCC(O)=O
Molecular Weight (g/mol) 390.46
Formula Weight 390.46
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.