missing translation for 'onlineSavingsMsg'
Learn More

LTURM34, MedChemExpress

Product Code. 16473488
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16473488

Brand: Medchem Express HY101667

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress LTURM34 is a specific DNA-PK inhibitor (IC50=34 nM). LTURM34 exhibits 170-fold selectivity for DNA-PK over PI3K. LTURM34 shows potent antiproliferative activity in a wide range of tumor cell lines.

LTURM34 is a specific DNA-PK inhibitor (IC50=34 nM). LTURM34 exhibits 170-fold selectivity for DNA-PK over PI3K. LTURM34 shows potent antiproliferative activity in a wide range of tumor cell lines.

Specifications

Chemical Name or Material LTURM34
CAS 1879887-96-3
Color White
Purity Grade Notes Research
Molecular Formula C24H18N2O3S
Quantity 10 mg
Solubility Information DMSO : 65 mg/mL (156.82 mM; Need ultrasonic)
SMILES O=C1C2=CC=CC(C3=CC=CC4=C3SC5=C4C=CC=C5)=C2OC(N6CCOCC6)=N1
Molecular Weight (g/mol) 414.48
Formula Weight 414.48
Percent Purity 99.89%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.