missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ LRP8 Recombinant Rabbit Monoclonal Antibody (23GB2520), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
217.00 € - 590.00 €
Specifications
| Antigen | LRP8 |
|---|---|
| Clone | 23GB2520 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088985
![]() |
Invitrogen™
MA553502 |
100 μL |
590.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089629
|
Invitrogen™
MA553503 |
20 μL |
217.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
LRP8 Recombinant Monoclonal antibody specifically detects LRP8 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| LRP8 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q14114, Q924X6 | |
| 16975, 362558, 7804 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB2520 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| 4932703M08Rik; AA921429; AI848122; ApoE receptor 2; APOER2; Apolipoprotein E receptor 2; HSZ75190; LDL receptor related protein 8; low density lipoprotein receptor relative, 8 ligand binding repeats, brain; low density lipoprotein receptor-related protein 8, apolipoprotein e receptor; low-density lipoprotein receptor-related protein 8; Lr8b; Lrp8; LRP-8; MCI1; Protein LR8B | |
| A synthesized peptide derived from human ApoER2 (900-963AA). | |
| Primary | |
| -20°C | |
| LRP8 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title