missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ LRP15 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA553323

Product Code. 15791261

  • 578.00 € / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: CPKGCLCSSS GGLNVTCSNA NLKEIPRDLP PETVLLYLDS NQITSIPNEI FKDLHQLRVL NLSKNGIEFI DEHAFKGVAE TLQTLDLSDN RIQSVHKNAF NNLKARARIA NNPWHCDCTL QQVLRSMASN HETAHNVICK TSVLD Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

The protein encoded by this gene is a tumor suppressor, with lowered expression levels found in gastric, renal, colorectal, lung, and breast cancer tissues. The promoter of this gene is frequently hypermethylated in these cancer tissues, although the hypermethylation does not appear to be the cause of the reduced expression of this gene. Several transcript variants encoding the same protein have been found for this gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

LRP15
Polyclonal
Unconjugated
LRRC3B
BC019794; leucine rich repeat containing 3B; leucine-rich repeat protein LRP15; Leucine-rich repeat-containing protein 3B; LRP15; LRRC3B; RGD1311897; UNQ195/PRO221
Rabbit
Antigen affinity chromatography
RUO
116135
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q96PB8
LRRC3B
Recombinant protein corresponding to Human LRP15. Recombinant protein control fragment (Product #RP-90526).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.