Learn More
LRFN5 Rabbit, anti-Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 26856-1-AP-20UL
Description
The LRFN5 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human LRFN5. This antibody recognizes mouse, rat antigen. The LRFN5 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| LRFN5 | |
| 350 μg/mL | |
| Rabbit | |
| Primary | |
| Mouse, Rat | |
| 150 kDa | |
| IgG |
| Western Blot, ELISA | |
| C14orf146, FIGLER8, LRFN5, SALM5 | |
| 20 μL | |
| 145581 | |
| Store at -20°C. Stable for one year after shipment. | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.