Learn More
LPL Monoclonal Antibody (OTI4G7), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503880
Description
LPL encodes lipoprotein lipase, which is expressed in heart, muscle, and adipose tissue. LPL functions as a homodimer, and has the dual functions of triglyceride hydrolase and ligand/bridging factor for receptor-mediated lipoprotein uptake. Severe mutations that cause LPL deficiency result in type I hyperlipoproteinemia, while less extreme mutations in LPL are linked to many disorders of lipoprotein metabolism.Specifications
| LPL | |
| Monoclonal | |
| 0.82 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P06858 | |
| LPL | |
| Human recombit protein fragment corresponding to amino acids 28-475 of human LPL produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI4G7 | |
| Unconjugated | |
| LPL | |
| HDLCQ11, LIPD | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 4023 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.