missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ LP 922056

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
LP 922056 is a notum pectinacetylesterase inhibitor. LP 922056 activates wnt signaling in vitro (EC50 values are 21 nM in human and 55 nM in mouse cell-based assay). It increases cortical bone thickness in mouse model of bone growth. LP 922056 also inhibits growth of Apc-mutant cell lines and proliferation of intestinal adenomas in vivo.
Specifications
Specifications
| Chemical Name or Material | 2-[(6-Chloro-7-cyclopropylthieno[3,2-d]pyrimidin-4-yl)thio]acetic acid |
| CAS | 1365060-22-5 |
| Quantity | 10 mg |
| Molecular Formula | C11H9ClN2O2S2 |
| Purity | 98% |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction