missing translation for 'onlineSavingsMsg'
Learn More

Loxapine succinate, MedChemExpress

Product Code. p-200096217
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16437276 100 mg 100mg
16411846 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16437276 Supplier Medchem Express Supplier No. HY17390A

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Loxapine succinate is an orally active dopamine inhibitor, 5-HT receptor antagonist and also a dibenzoxazepine anti-psychotic agent.

Loxapine succinate is an orally active dopamine inhibitor, 5-HT receptor antagonist and also a dibenzoxazepine anti-psychotic agent.

Specifications

Chemical Name or Material Loxapine succinate
CAS 27833-64-3
Color Off-White
Purity Grade Notes Research
Molecular Formula C22H24ClN3O5
Quantity 100 mg
Solubility Information DMSO : 100 mg/mL (224.27 mM; Need ultrasonic) ∣H2O : 2.5 mg/mL (5.61 mM; ultrasonic and warming and heat to 60°C)
SMILES O=C(O)CCC(O)=O.CN1CCN(C2=NC3=CC=CC=C3OC4=CC=C(Cl)C=C24)CC1
Molecular Weight (g/mol) 445.9
Formula Weight 445.9
Percent Purity 99.81%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.