Learn More
Invitrogen™ LOX-1 Monoclonal Antibody (23C11), FITC

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA549085
Description
Monoclonal antibody 23C11 neutralizes LOX-1 and inhibits Hsp70 binding to dendritic cells and Hsp70-induced antigen cross-presentation. In vivo, targeting LOX-1 with a tumor antigen using anti-LOX-1 antibody 23C11 induces anti-tumor immunity.
LOX-1 (lectin-like oxidized LDL receptor -1) is a 40-50 kDa type II membrane glycoprotein class E scavenger receptor expressed by endothelial cells, macrophages, activated platelets and smooth muscle cells. LOX-1 binds to a wide range of ligands, including oxidized low-density lipoprotein (oxLDL), hypochlorite modified high-density lipoprotein (HDL), aged /apoptotic cells, activated platelets and bacteria, reflecting its versatile physiological functions.
Specifications
| LOX-1 | |
| Monoclonal | |
| 100 μg/mL | |
| PBS with 1% BSA and 0.02% sodium azide | |
| P78380 | |
| OLR1 | |
| Fusion protein of extracellular domain (aa 71-273) of human LOX-1 with murine Fc-gamma1 (LOX-1-muFc) produced in PEAK cells. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Flow Cytometry, Functional Assay, Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 23C11 | |
| FITC | |
| OLR1 | |
| CLEC8A; C-type lectin domain family 8 member A; hLOX 1; hLOX1; hLOX-1; lectin-like oxidized LDL receptor; lectin-like oxidized LDL receptor 1; lectin-like oxidized low-density lipoprotein receptor; Lectin-like oxidized low-density lipoprotein receptor-1; lectin-like oxLDL receptor 1; lectin-type oxidized LDL receptor 1; LOX 1; LOX1; LOX-1; LOXIN; Oldlr1; Oldr1; OLR1; ox LDL receptor 1; oxidised low density lipoprotein (lectin-like) receptor 1; oxidized low density lipoprotein (lectin-like) receptor 1; oxidized low density lipoprotein receptor 1; oxidized low-density lipoprotein receptor 1; Oxidized low-density lipoprotein receptor 1, soluble form; ox-LDL receptor 1; Scare1; scavenger receptor class E, member 1; sLOX 1; SLOX1; soluble LOX 1; SR-EI | |
| Mouse | |
| Protein G | |
| RUO | |
| 4973 | |
| 4°C, store in dark | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.