Learn More
Greiner Bio-One Low Density Polyethylene Serum Pipettes
GREENER_CHOICE
Product Code. p-7075384
Click to view available options
Sterility:
Non-sterile
Sterile
Packaging Type:
25/bag
500/bag
Unit Size:
Pack of 1000
Pack of 1500
Product Code. 10357941
Brand: Greiner Bio-One 612301
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Greiner Bio-One Serum Pipettes are ideally suited for rapid transfer of liquids.
  • With graduation
  • Material: Low density polyethylene
  • Frapid transfer of liquids
  • For single use only
TRUSTED_SUSTAINABILITY
Specifications
Material LDPE
Sterility Non-sterile
For Use With (Application) Rapid transfer of liquids
Graduated Yes
Quantity 1500/Cs.
Packaging Type 500/bag
Volume (Metric) 2.5 mL
Length (Metric) 153 mm
Type Serum Pipette