Learn More
Lovibond™ Gardner Scale 3000 Comparator Accessories
Product Code. 10139611
Change view
Click to view available options
Type:
Test Tube
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Type Unit Size
10139611 Test Tube Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10139611 Supplier Lovibond™ Supplier No. 352280
Estimated Shipment: 15-01-2027
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Test tubes, 10.65mm diameter, accessories for Gardner Comparator
For use with the Lovibond Gardner Scale 3000 Comparator (09-053-409).
Specifications
Type Test Tube
For Use With (Equipment) Gardner comparator 3000 and FAC comparator 3000