Learn More
Lovastatin Hydroxy Acid, Sodium Salt, TRC
Specifications
Specifications
| Chemical Name or Material | (3R,5R)-7-[(1S,2S,6R,8S,8aR)-2,6-Dimethyl-8-[((2S)-2-methylbutanoyl]oxy)-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoic Acid Sodium Salt (Lovastatin Hydroxy Acid Sodium Salt) |
| CAS | 75225-50-2 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C24 H37 O6 . Na |
| Quantity | 100 mg |
| Synonym | (3R,5R)-7-[(1S,2S,6R,8S,8aR)-2,6-Dimethyl-8-[[(2S)-2-methylbutanoyl]oxy]-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoic Acid Sodium Salt, Lovastatin Imp. B (EP) as Sodium Salt, Lovastatin Hydroxy Acid Sodium Salt, 1-Naphthaleneheptanoic acid, 1,2,6,7,8,8a-hexahydro-β,Δ-dihydroxy-2,6-dimethyl-8-(2-methyl-1-oxobutoxy)-, sodium salt (1:1), (βR,ΔR,1S,2S,6R,8S,8aR)-, 1-Naphthaleneheptanoic acid, 1,2,6,7,8,8a-hexahydro-β,Δ-dihydroxy-2,6-dimethyl-8-(2-methyl-1-oxobutoxy)-, monosodium salt, [1S-[1α(βS*,ΔS*),2α,6β,8β(R*),8aα]]-, 1-Naphthaleneheptanoic acid, 1,2,6,7,8,8a-hexahydro-β,Δ-dihydroxy-2,6-dimethyl-8-[(2S)-2-methyl-1-oxobutoxy]-, monosodium salt, (βR,ΔR,1S,2S,6R,8S,8aR)- (9CI), Lovastatin sodium salt, MB 530B, MSD 803, sodium salt, Monacolin K sodium salt, Sodium mevinolinate |
| InChI Formula | InChI=1S/C24H38O6.Na/c1-5-15(3)24(29)30-21-11-14(2)10-17-7-6-16(4)20(23(17)21)9-8-18(25)12-19(26)13-22(27)28;/h6-7,10,14-16,18-21,23,25-26H,5,8-9,11-13H2,1-4H3,(H,27,28);/q;+1/p-1/t14-,15-,16-,18+,19+,20-,21-,23-;/m0./s1 |
| SMILES | [Na+].CC[C@H](C)C(=O)O[C@H]1C[C@@H](C)C=C2C=C[C@H](C)[C@H](CC[C@@H](O)C[C@@H](O)CC(=O)[O-])[C@@H]12 |
| IUPAC Name | sodium;(3R,5R)-7-[(1S,2S,6R,8S,8aR)-2,6-dimethyl-8-[(2S)-2-methylbutanoyl]oxy-1,2,6,7,8,8a-hexahydronaphthalen-1-yl]-3,5-dihydroxyheptanoate |
| Molecular Weight (g/mol) | 444.54 |
| Show More |