Learn More
Lorglumide sodium salt, MedChemExpress
Product Code. 16408708
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
Unit Size:
10mg
5mg
Product Code. 16408708
Brand: Medchem Express HYB1439B
Please to purchase this item. Need a web account? Register with us today!
MedChemExpress Lorglumide sodium salt (CR-1409 sodium salt) is a potent cholecystokinin (CCK) receptor antagonist.
Lorglumide sodium salt (CR-1409 sodium salt) is a potent cholecystokinin (CCK) receptor antagonist.
Specifications
Chemical Name or Material Lorglumide sodium salt
CAS 1021868-76-7
Color White
Purity Grade Notes Research
Molecular Formula C22H31Cl2N2NaO4
Quantity 10 mg
Synonym CR-1409 sodium salt
Solubility Information H2O : ≥ 100 mg/mL (207.73 mM) ∣DMSO : 100 mg/mL (207.73 mM; Need ultrasonic)
SMILES ClC1=C(C=CC(C(NC(CCC(O[Na])=O)C(N(CCCCC)CCCCC)=O)=O)=C1)Cl
Molecular Weight (g/mol) 481.39
Formula Weight 481.39
Percent Purity 99.0%
Grade Research
Physical Form Powder
For Use With (Application) Metabolism-protein/nucleotide metabolism